YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 5 µm, Semi-Preparative/Preparative, YMC America
Supplier: YMC America,Inc
TS12S05-1020WX TS12S05-1030WX TS12S05-1553DX TS12S05-2520WX TS12S05-2530WX TS12S05-2553DX TS12S05-L530WX TS12S05-1053DX TS12S05-1520WX TS12S05-1530WX
76027-744EA 7524.69 CAD
76027-744 76027-746 76027-750 76027-752 76027-754 76027-758 76027-760 76027-788 76027-790 76027-792
YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 5 µm, Semi-Preparative/Preparative, YMC America
Chromatography Columns
YMC-Triart Silica columns are intended for use in normal phase and HILIC chromatography modes.
  • Bare organic/inorganic hybrid silica particle
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • May be used in normal phase or HILIC chromatography modes
YMC-Triart Silica is available in columns packed with particle sizes of 1.9, 3, 5, 10, 15, and 20 µm, intended for use on both SFC and HPLC systems.
Columns are available in many sizes. Please inquire if you do not see the column geometry you desire.