Steam Generator
Supplier: EISCO SCIENTIFIC LLC
PH0467SG
470230-186EA 72.9 CAD
470230-186
Steam Generator
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Ideal for labs to generate heat and pressure for water vapor.
  • Volume held is typically enough for 2 stations for 1 lab period, place cold water in AL reservoir introduce hot element using sample carrier and steam is produced
  • General use for a this steam generator is for creating high pressure and gas for physics and chemistry such as thermal expansion (linear rod expansion) type experiments
  • Works well with labs PH0371B and PH0371A
  • Caution: Creating steam is a dangerous process and creates high temperature gases and pressure, this product is for scientific lab use only
The steam generator is intended for scientific laboratories looking to generate heat and pressure for water vapor. General use for a this steam generator is for creating high pressure and gas for physics and chemistry such as thermal expansion (linear rod expansion) type experiments. Caution: Creating steam is a dangerous process and creates high temperature gases and pressure, this product is for scientific lab use only.
Ordering information: Apparatus contains (1) Thick Wall Aluminum can - Approximately 1 L with nozzle for steam outlet, (1) AL Sample carrier, (1) Spring Handle, (1) Tripod Cast Base, (1) Size 13 Stopper, (1) Size 13 Stopper with hole for thermometer.
Caution: Creating steam is a dangerous process and creates high temperature gases and pressure; this product is for scientific lab use only.