Hydrochloric acid 5%, Assurance® Grade for calibration blank in ICP, for atomic absorption spectroscopy
Supplier: SPEX CERTIPREP LLC
Synonyms:
Muriatic acid
, Hydrochloric acid solution
PLBLK-HCL
SXPLBLK-HCLEA
143.02
CAD
SXPLBLK-HCL
Hydrochloric acid 5%, Assurance® Grade for calibration blank in ICP, for atomic absorption spectroscopy
Hydrochloric acid
Calibration and matrix blank.
May be used to dilute your multi-element standards or can be run directly as a blank to establish your base line. Do not use any acid or water as a diluent if you are not certain of its purity.
Formula:
HCl MW: 36.46 g/mol |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...