VWR® Burets with Straight Bore PTFE Stopcock, Class A, Serialized
Supplier: VWR International
75997-906 75997-908 75997-910 75997-912
75997-906CS 548.88 CAD
75997-906 75997-908 75997-910 75997-912
VWR® Burets with Straight Bore PTFE Stopcock, Class A, Serialized
Burettes
VWR® Burets are made from Borosilicate 3.3 glass, ASTM 438 Type 1, Class A standard
  • Supplied with a chemically-resistant, self-lubricating PTFE stopcock plug
  • Used primarily for titration, to deliver one reactant until the precise endpoint is reached
  • Calibration and tolerances per ASTM E287, ASTM E542, ASTM E694, and ISO 385 Standards
The graduations are typically calibrated "to deliver" (marked "TD" or "EX") at 20 to 27 °C and indicated correspondingly on a label. Serialized, individually certified products.
These Burets are calibrated to deliver a precise volume at a particular temperature.