VWR® Weighing Bottles with Stopper, Tall Form, Cylindrical
Supplier: VWR International
75997-898 75997-900 75997-902 75998-016 75998-018 75998-020
75997-898CS 844.66 CAD
75997-898 75997-900 75997-902 75998-016 75998-018 75998-020
VWR® Weighing Bottles with Stopper, Tall Form, Cylindrical
Bottles Weighing Bottles
These tall form weighing bottles are used for precise weighing of solids.
  • Complies ASTM – 438, TYPE-1, Boro 3.3 glass
  • These bottles are chemically resistant and mechanically strong
These weighing bottles come with a hollow style plug type glass stopper which seals the contents to prevent spilling.
Manufactured from 3.3 expansion ASTM 438 Type 1 Class A standard borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable