Pierce™ Surfact-Amps™ Detergent Solutions, Thermo Scientific
Supplier: Thermo Scientific
28314 28320 28324 28328 28332 28230 28313 28316 28321 28329 28336 85111 85112 85113 85114 85115 85117 85118 85124
CAPI28314EA 403.24 CAD
CAPI28314 CAPI28320 CAPI28324 CAPI28328 CAPI28332 CAPI28230 CAPI28313 CAPI28316 CAPI28321 CAPI28329 CAPI28336 CAPI85111 CAPI85112 CAPI85113 CAPI85114 CAPI85115 CAPI85117 CAPI85118 CAPI85124
Pierce™ Surfact-Amps™ Detergent Solutions, Thermo Scientific
Protein Extraction Protein Extraction Reagents
Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques.
  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life
  • Pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products
Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilize membrane proteins and lipids, control protein crystallization and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.
Order Now
SPECIFICATIONS
Environmentally Preferable