Ward’s® Automatic Autoclave Series
Supplier: VWR International
470230-596 470230-598 B4000-28-120
470230-596EA 9720.8 CAD
470230-596 470230-598 470351-285
Ward’s® Automatic Autoclave Series
Sterilizers Autoclaves (Steam Sterilizers)
Three convenient options for benchtop sterilization.
  • Rapid, complete sterilization for educational/research use
  • Built-in water tank doesn't require external water supply
  • Safety interlocks prevent door from opening while pressurized
  • Stainless steel sterilization chambers
  • One-year warranty
Ward's® BioClaves offer three preset standard cycle options, for sterilization of liquids, wrapped instruments or unwrapped instruments/plastic ware. A fourth option "Dry Only" is also available for additional drying time to be added to the end of a cycle.
Order Now
SPECIFICATIONS
Environmentally Preferable