Density Set, Delrin, Set 12

Supplier: Wards
PH0113K
470230-050EA 50.4 CAD
470230-050
Density Set, Delrin, Set 12
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Set of 12 Delrin cylindrical bars of different length but the same diameter.

  • Great for classroom use
  • High quality wooden storage block
  • Durable and economically friendly materials

Students learn that the density of a substance is constant even as its mass and volume change. Comes in an attractive wooden holder.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR