Metal shapes, Set of 12
Supplier: Wards
PH0113
470230-046EA 81.55 CAD
470230-046
Metal shapes, Set of 12
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
For density determinations, set of twelve in assorted sizes, shapes and masses.
  • 12 objects in assorted shapes, sizes and masses
  • Brass, iron, aluminum, copper, zinc, lead
  • Mass range of 12 - 25 grams
The Labs Set of Metal Blocks is used for density determinations. It includes two of each brass, iron, aluminum, copper, zinc and lead. Masses vary from 12-25 grams.