Density Cubes with Hooks

Supplier: EISCO SCIENTIFIC LLC
PH0108R
470230-036EA 103.8 CAD
470230-036
Density Cubes with Hooks
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Density Cubes, Set of 7 Metals with Hooks, (20 mm) sides.

  • These density cubes measure 0.75" cubed with hooks in each cube.
  • Each cube is marked for identification.
  • Set includes the following metals Fe, Cu, Al, Br, Sn, Zn, Pb.
  • Comes with plastic case to contain all cubes for ease of storage and organization.

The set consists of seven 20 mm cubes and is perfect for investigating density.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR