Specific Gravity Cubes
Supplier: Wards
PH0108
PH0108A
PH0108D
PH0108E
PH0108F
470230-020EA
46
CAD
470230-020
470230-022
470230-024
470230-026
470230-028
Specific Gravity Cubes
Educational Materials
Physics Educational Materials
Properties of Matter Learning Activities in Physics
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...