VWR® Volumetric Flask, Clear Glass, with Red Vertical Stripe and Glass Penny Head Stopper, Class A
Supplier: VWR International
134.237.08 131.237.09 131.237.10 131.237.11 134.237.04 134.237.05 134.237.06 134.237.07
76008-580CS 659.33 CAD
76008-580 76008-582 76008-584 76008-586 76008-700 76008-702 76008-704 76008-706
VWR® Volumetric Flask, Clear Glass, with Red Vertical Stripe and Glass Penny Head Stopper, Class A
Flasks Volumetric Flasks
Flasks are lot certified and supplied with a glass penny head stopper
  • Made from Borosilicate 3.3 expansion glass
  • Calibrated 'to Contain'
  • Batch Certificate included
The red stripe makes it easier to see the meniscus.