LB Minitube Preps with Kanamycin, Sterile, Teknova
Supplier: TEKNOVA CA
L8530 L8535
CA100220-212EA 575.72 CAD
CA100220-212 CA100220-214
LB Minitube Preps with Kanamycin, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For plasmid DNA minipreps
  • Each tube contains 3 mL of broth
  • Pack of 25 tubes in a reusable rack
LB is packaged in snap-cap culture tubes that can be used to both grow cells and centrifuge them once they are grown.