LB Minitube Preps with Ampicillin, Sterile, Teknova
Supplier: TEKNOVA CA
L8505 L8510
CA100220-202EA 581.55 CAD
CA100220-202 CA100220-204
LB Minitube Preps with Ampicillin, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For plasmid DNA minipreps
  • Each tube contains 3 mL of broth
  • Pack of 25 tubes in a reusable rack
LB is packaged in snap-cap culture tubes that can be used to both grow cells and centrifuge them once they are grown.