VWR® Erlenmeyer Flask with Standard Taper Outer Joint
Supplier: VWR International
76008-690 76008-692 76008-694 76008-696 76008-698
76008-690CS 315.15 CAD
76008-690 76008-692 76008-694 76008-696 76008-698
VWR® Erlenmeyer Flask with Standard Taper Outer Joint
Flasks Erlenmeyer Flasks
High-quality durable flask featuring a ground joint and uniform wall thickness and are a great value.
  • Manufactured from ASTM E438 Type 1
  • Class A borosilicate 3.3 expansion glass
VWR® Erlenmeyer flasks are used to measure, mix or hold fluids or chemicals commonly used in laboratories.