VWR® Round Bottom Flask, 3 Necks, Parallel
Supplier: VWR International
76008-650 76008-652 76008-654 76008-656 76008-658 76008-660 76008-662 76008-664 76008-666 76008-668 76008-670 76008-672 76008-674 76008-676 76008-678 76008-680 76008-682 76008-684 76008-686 76008-688
76008-650CS 276.41 CAD
76008-650 76008-652 76008-654 76008-656 76008-658 76008-660 76008-662 76008-664 76008-666 76008-668 76008-670 76008-672 76008-674 76008-676 76008-678 76008-680 76008-682 76008-684 76008-686 76008-688
VWR® Round Bottom Flask, 3 Necks, Parallel
Flasks Round Bottom Flasks
Durable round bottom flasks with 3 parallel necks.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.
Order Now
SPECIFICATIONS
Environmentally Preferable