Super Broth Dehydrated Media, Teknova
Supplier: TEKNOVA CA
S0140 S0145 S0135
CA100217-180EA 872.16 CAD
CA100217-180 CA100218-072 CA100219-646 CA100219-876
Super Broth Dehydrated Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For use in propagation of bacteriophage lamda.
  • Multiple sizes available
Contains 3.2% tryptone, 2.0% yeast extract, 0.5% sodium chloride, 0.99% Potassium Phosphate, Dibasic.
Ambient shipping conditions. Store at room temperature.