J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKRD012-00 BAKRD012-02 BAKRD014-00 BAKRD014-02 BAKRD014-04 BAKRD015-04
CAJTD012-0CS 1267.87 CAD
CAJTD012-0 CAJTD012-2 CAJTD012-4 CAJTD014-0 CAJTD014-2 CAJTD014-4 CAJTD015-4
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Buffers Biological Buffers
DILUT-IT™ is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities.