LB Lennox Broth Dehydrated Media, Teknova
Supplier: TEKNOVA CA
L9305 L9320
CA100219-286EA 364.27 CAD
CA100219-286 CA200066-630
LB Lennox Broth Dehydrated Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of E. coli and other enteric bacteria.
  • Add 20 grams to 1 Liter of water to reconstitute
  • Autoclave to sterilize
LB Lennox contains less salt than the most common formulation of LB, known as LB Miller
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 0.5% NaCl (5.0g/L).
Ambient shipping conditions. Store at room temperature.
LB Lennox is commonly used for cloning in recombinant E. coli and selection with salt-sensitive antibiotics such as Blasticidin, CloNat, Hygromycin, Puromycin and Zeocin.