LB Broth Dehydrated Media, Teknova
Supplier: TEKNOVA CA
L9140 L9145 L9135 L9130
CA100218-046EA 957.67 CAD
CA100218-046 CA100218-394 CA100218-844 CA100219-108
LB Broth Dehydrated Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of E. coli and other enteric bacteria. LB Miller is the most commonly used formulation, often referred to as LB.
  • Use 25 grams per Liter to reconstitute the LB Broth
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 1.0% NaCl (10.0g/L).
Ambient shipping conditions. Store at room temperature.