LB Agar Dehydrated Media, Teknova
Supplier: TEKNOVA CA
L9105 L9110 L9115 L9120
CA100216-806EA 532.79 CAD
CA100216-806 CA100217-750 CA100217-986 CA100218-380
LB Agar Dehydrated Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of E. coli and other enteric bacteria.
  • Use 40 grams per Liter to reconstitute the LB agar
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 1.0% NaCl (10.0g/L), 1.5% Agar (15.0g/L).
Ambient shipping conditions. Store at room temperature.