Arabinose Agar Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M1576-2.5KG M1576-5KG
CA89224-284EA 4423.65 CAD
CA89224-284 CA89224-286
Arabinose Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended to differentiate enterococcus faecium from Enterococcus faecalis.
  • Multiple sizes available