NZYM Plus Broth, Teknova
Supplier: TEKNOVA CA
N1225 N1225
CA100218-018PK 503.5 CAD
100218-018 CA100218-018
NZYM Plus Broth, Teknova
Media for Microbiology Prepared Media for Microbiology
Used for the maintenance and propagation of bacteriophage lambda.
  • 10 tubes per pack with 10mL broth per tube
  • Sterile
Contains 1.0% Casein digest (NZ amine), 0.5% NaCl, 0.5% yeast extract, 0.4% glucose, 12.5 mm MgSO4.7H2O, and 12.5 mm MgCl2.
Ambient shipping conditions. Store at room temperature.