Tryptic Soy Agar Plates, Teknova
Supplier: Teknova
T0144 T0144 T0144
CA100217-300PK 377.26 CAD
100217-300 CA100217-300 77442-968
Tryptic Soy Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
For isolating and cultivating a variety of fastidious microorganisms.
  • 20 Plates
  • Sterile
Contains 1.5% Tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar.
Ambient shipping conditions. Store at room temperature.