Photostability Test Chambers, 7540 Series, Caron Products
Supplier: Caron Products
7540-11-1 7540-11-2 7540-11-3 7545-11-1 7545-11-2 7545-11-3
76102-342EA 110833.59 CAD
76102-342 76102-344 76102-346 76102-348 76102-350 76102-352
Photostability Test Chambers, 7540 Series, Caron Products
Testing Chambers Photostability Chambers
With full visible and UVA ICH Q1B exposures in as little as 42 hours, the 7450 series provides results fast. Many competing chambers take 2 to 3 times as long. Fast photoexposure allows users to test more samples, reducing drug development time, and saving money.
  • Highly reflective specular aluminum interior panels and separate UV and visible light banks create edge-to-edge light uniformity, with over 75% of shelf space usable
  • Separate UV and visible Q1B Option II bulbs provide speed without risking exposure overshoot
  • Tight temperature uniformity prevents
More Product Information