VWR® Tube Top Vacuum Filters

Supplier: VWR International
76012-772 76012-774 76012-776 76012-778 76022-490
76012-772CS 252.43 CAD
76012-772 76012-774 76012-776 76012-778 76022-490
VWR® Tube Top Vacuum Filters
Filters Bottle Top Filters
Minimize unnecessary transfers by filtering directly into 50 ml centrifuge tubes.

  • Tube top vacuum filters
  • 50 mm Ø membrane
  • Two pore sizes: 0.2 and 0.45 μm
  • Gamma radiation sterilized
  • Non-pyrogenic

PES membranes offer low protein binding and low extractables.

The 0.2 µm pore size is ideal for sterilization applications and media preparation. The 0.45 µm pore size is ideal for buffer clarification.

Ideal for tissue culture applications.

Certifications: ISO13485 :2003 and ISO9001:2008.

Accessories information: The Tube Top Filter Stand features 7 openings, suitable for holding 1.5 and 2.0 ml microcentrifuge tubes, or 15 and 50 ml conical centrifuge tubes. It provides an operating temperature range of –80 to 121 °C, and may be autoclaved.

Delivery information: Each polypropylene centrifuge tube is supplied with an individually wrapped cap for storage.

Packaging: Packaged in resealable bags. Each individual unit is lot-numbered for easy identification and tracking.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR