Outdoor Weather-Resistant Polymer Label, Silver, 3" Core, Brady
Supplier: Brady
148787 148784 148788 148782 148781 148785 148786 148789 148790
76000-840EA 618 CAD
76000-848 76000-838 76000-881 76000-882 76000-840 76000-842 76000-844 76000-846 76000-836
Outdoor Weather-Resistant Polymer Label, Silver, 3" Core, Brady
Labels
The B-8591 material is an extremely durable, print-on-demand thermal transfer label solution that resists UV, abrasion, and fluids in outdoor use.
  • 12+ year outdoor durability reduces identification failures and the costs associated with losing this information
  • No over-laminate means durability is drastically improved, without adding an over-laminate over the print
  • Quick and easy application with pressure-sensitive adhesive
Certifications: Meets regulatory specifications for halogen-free applications.