LB Agar Plates with 50 µg/ml Kanamycin, X-gal and IPTG, Teknova
Supplier: TEKNOVA CA
L5909
L1909
CA100217-686PK
530.41
CAD
CA100217-686
CA100219-332
LB Agar Plates with 50 µg/ml Kanamycin, X-gal and IPTG, Teknova
Media for Microbiology
Prepared Media for Microbiology
For use with kanamycin resistant strains and strains harboring plasmids such as pPIC9K, pCR®-Blunt, pUC9P series plasmids.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50 µg/mL Kanamycin, 60 µg/mL X-Gal, 0.1 mM IPTG
Ambient shipping conditions. Refrigerate to store.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...