LB Agar Plates with 100 µg/ml Carbenicillin, X-gal and IPTG, Teknova
Supplier: TEKNOVA CA
L5906 L1906
CA100217-680PK 503.51 CAD
CA100217-680 CA100218-312 CA100219-326
LB Agar Plates with 100 µg/ml Carbenicillin, X-gal and IPTG, Teknova
Media for Microbiology Prepared Media for Microbiology
Carbenicillin is an analog of ampicillin. For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 100 µg/ml carbenicillin, 0.1 mM IPTG, 60 µg/ml X-gal.
Ambient shipping conditions. Refrigerate to store.