LB Agar Plates with 50 µg/ml Carbenicillin, X gal and IPTG, Teknova
Supplier: TEKNOVA CA
L3904
L1904
CA100217-636PK
1316.76
CAD
CA100217-636
CA100219-322
LB Agar Plates with 50 µg/ml Carbenicillin, X gal and IPTG, Teknova
Media for Microbiology
Prepared Media for Microbiology
Carbenicillin is an analog of ampicillin. For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 50µg/ml carbenicillin, 0.1 mM IPTG, 60 µg/ml X-gal.
Ambient shipping conditions. Refrigerate to store.