LB Agar Plates with 150 µg/ml Carbenicillin, 34 µg/ml Chloramphenicol, 50 µg/mL Kanamycin and 10 µg/ml Tetracycline, Teknova
Supplier: TEKNOVA CA
L1405
CA200066-676PK 909.57 CAD
CA200066-676
LB Agar Plates with 150 µg/ml Carbenicillin, 34 µg/ml Chloramphenicol, 50 µg/mL Kanamycin and 10 µg/ml Tetracycline, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with carbenicillin, chloramphenicol kanamycin, and tetracycline resistant strains.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 150 µg/ml carbenicillin, 34 µg/ml chloramphenicol, 50 µg/ml kanamycin, and 10 µg/ml tetracycline.
Ambient shipping conditions. Refrigerate to store.