LB Lennox Agar Plates with Zeocin 100, Teknova
Supplier: Teknova
L1039
CA100219-832PK 2312.9 CAD
CA100219-832
LB Lennox Agar Plates with Zeocin 100, Teknova
Media for Microbiology Prepared Media for Microbiology
LB lennox agar plates with zeocin 100.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 100 µg/mL Zeocin.
Ambient shipping conditions. Refrigerate to store.