LB Lennox Agar Plates with Zeocin 50, Teknova
Supplier: Teknova
L1038
CA100219-966PK 1905.75 CAD
CA100219-966
LB Lennox Agar Plates with Zeocin 50, Teknova
Media for Microbiology Prepared Media for Microbiology
LB lennox agar plates with zeocin 50.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% yeast extract, 0.5% sodium chloride, 2.0% agar, 50 µg/mL zeocin.
Ambient shipping conditions. Refrigerate to store.