LB Agar Plates with Spectinomycin 100, Teknova
Supplier: TEKNOVA CA
L1028
L5028
CA100216-594PK
322.45
CAD
CA100216-594
CA100217-260
LB Agar Plates with Spectinomycin 100, Teknova
Media for Microbiology
Prepared Media for Microbiology
For use with spectinomycin resistant strains and strains harboring plasmids such as pGV1122, pUCD4, pGV1106 series plasmids.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 100 µg/ml spectinomycin. It is used with spectinomycin resistant strains and strains harboring plasmids such as pGV1122, pUCD4, pGV1106 series plasmids.
Ambient shipping conditions. Refrigerate to store.