LB Agar Plates with 50 µg/ml Kanamycin, Teknova
Supplier: TEKNOVA CA
L1025
L5025
L3025
L2025
CA100216-588PK
572.04
CAD
CA100216-588
CA100217-254
CA100218-230
CA100218-444
LB Agar Plates with 50 µg/ml Kanamycin, Teknova
Media for Microbiology
Prepared Media for Microbiology
For growing strains of recombinant E. coli transformed with plasmids containing the resistance gene for Kanamycin (kan).
- Sleeve of 20 plates
- Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% Sodium chloride (NaCl) (10.0 g/L), 1.5% Agar (15.0 g/L), 50 µg/mL Kanamycin.
Ambient shipping conditions. Store at 2 to 8 °C.