LB Agar Plates with 25 µg/ml Kanamycin, Teknova
Supplier: Teknova
L5023 L1023 L5623 L1023 L1023 L3023 L5623 L5023 L3023
CA100218-226PK 1297.16 CAD
101414-080 100216-584 CA100218-226 CA100216-584 CA100217-250 100217-250 100218-226 77443-350 CA101414-080
LB Agar Plates with 25 µg/ml Kanamycin, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with kanamycin resistant strains and strains transformed with plasmids such as pPIC9K, TOPO-TA, TOPO pCR-Blunt, and pUC9P.
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract, (5.0 g/L), 1.0% Sodium Chloride (10.0 g/L), 1.5% Agar (15.0 g/L), 25 µg/ml Kanamycin.
Room temperature shipping conditions. Store at 2 to 8 °C.