LB Agar Plates with Chloramphenicol, Teknova
Supplier: TEKNOVA CA
L1015
L1016
CA100216-570PK
806.95
CAD
CA100216-570
CA100216-572
LB Agar Plates with Chloramphenicol, Teknova
Media for Microbiology
Prepared Media for Microbiology
For use with chloramphenicol resistant strains and strains harboring plasmids such as pBC KS+/-, pJAK17, pMMB208 series plasmids.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar chloramphenicol.
Ambient shipping conditions. Refrigerate to store.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...