LB Agar Plates with Chloramphenicol 12.5, Teknova

Supplier: Teknova
L1013 L1013
CA100216-566PK 804 CAD
100216-566 CA100216-566
LB Agar Plates with Chloramphenicol 12.5, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with chloramphenicol resistant strains and strains harboring plasmids such as pBC KS+/-, pJAK17, pMMB208 series plasmids.

  • Sleeve of 20 plates
  • Sterile

Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 12.5 µg/ml chloramphenicol.

Ambient shipping conditions. Refrigerate to store.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR