LB Agar Plates with 100 µg/ml Carbenicillin, Teknova
Supplier: TEKNOVA CA
L1010
L5010
L3010
L2010
CA100216-562PK
219.86
CAD
CA100216-562
CA100217-226
CA100218-202
CA100218-414
LB Agar Plates with 100 µg/ml Carbenicillin, Teknova
Media for Microbiology
Prepared Media for Microbiology
Carbenicillin is an analog of ampicillin
- Sleeve of 20 plates
- Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% sodium chloride (NaCl) (10.0 g/L), 1.5% Agar (15.0 g/L), 100 µg/mL Carbenicillin.
Ambient shipping conditions. Store at 2 to 8 °C.
For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.