LB Agar Plates with 50 µg/ml Carbenicillin, Teknova
Supplier: TEKNOVA CA
L1008 L5008 L3008 L2008
CA100216-558PK 515.92 CAD
CA100216-558 CA100217-222 CA100218-198 CA100218-410
LB Agar Plates with 50 µg/ml Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
Carbenicillin is an analog of ampicillin. For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 50 µg/ml carbenicillin.
Ambient shipping conditions. Refrigerate to store.
Order Now
SPECIFICATIONS
Environmentally Preferable