LB Agar Plates with 100 µg/ml Ampicillin, Teknova
Supplier: TEKNOVA CA
L1004
L5004
L2004
CA100216-550PK
309.51
CAD
CA100216-550
CA100217-214
CA100218-402
LB Agar Plates with 100 µg/ml Ampicillin, Teknova
Media for Microbiology
Prepared Media for Microbiology
For use with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin (bla or ampR) such as pBluescript, pGEM, or the pUC series of plasmids.
- Sleeve of 20 plates
- Sterile
Ingredients: 1.0% tryptone (10.0 g/l), 0.5% yeast extract (5.0 g/l), 1.0% sodium chloride (NaCl) (10.0 g/l), 1.5% agar (15.0 g/l), 100 µg/ml ampicillin.
Ambient shipping conditions. Store at 2 to 8 °C.