LB Agar Plates with 50 µg/mL Ampicillin, Teknova
Supplier: TEKNOVA CA
L1002
L5002
CA100216-546PK
804
CAD
CA100216-546
CA100217-210
LB Agar Plates with 50 µg/mL Ampicillin, Teknova
Media for Microbiology
Prepared Media for Microbiology
For use with recombinant E. coli strains harboring plasmids such as pBluescript, pGEM, and the pUC series.
- 20 plates
- Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract, (5.0 g/L), 1.0% Sodium Chloride (10.0 g/L), 1.5% Agar (15.0 g/L), 50 µg/ml Ampicillin.
Room temperature shipping conditions. Store at 2 to 8 °C.