FastGene Cell Culture Protective Trays, Bulldog Bio
Supplier: Bulldog Bio
FGCC01F FGCC01
103370-366EA 252.31 CAD
103370-366 103370-368
FastGene Cell Culture Protective Trays, Bulldog Bio
Culture Dishes Cell Culture Dishes
Protect your valuable cells under the hood, in the incubator, and everywhere in between.
  • Works for plates, dishes, and flasks
  • Autoclavable
  • Replaceable <0.4 µm filter that lets air in and keeps pathogens out
Many cell biologists have had the frustrating experience of dealing with microbial infection or contamination by other cell lines. This problem can become quite severe when difficult-to-obtain primary or stem cells are affected. Once a culture container is infected, it can cause the infection to spread to other cell culture vessels throughout the lab. With the FastGene Cell Culture Protective Tray, contamination is now a problem of an earlier generation. These simple-to-use chambers provide a sterile and protective environment for your culture plates, dishes, and flasks in the incubator, cell culture hood, and elsewhere. Stop the spread of infection today!