Calcium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Supplier: SPEX CERTIPREP CA
PLCA2-2Y PLCA2-3Y PLCA2-3X PLCA2-2T PLCA2-2X PLCA2-3T
CA11020-444EA 306.54 CAD
CA11020-444 CA11023-474 CA11023-476 CA11023-480 CA11023-482 CA11033-296
Calcium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Standards Cationic Standards Calcium Standards
  • Traceable to NIST
  • Uncertainty of standards is reported on the Certificate of Analysis
  • Manufactured by experienced chemists to ensure highest quality standards
Caution: 1000 µg/ml = 1000 ppm.