Glass Baffles for Process Reactors, Chemglass
Supplier: Chemglass
CG-1971-120 CG-1971-122 CG-1971-132 CG-1971-136 CG-1971-70 CG-1971-72 CG-1971-74 CG-1971-66
16003-098EA 1628.26 CAD
16003-098 16003-100 16003-104 16003-106 16003-116 16003-118 16003-120 33501-378
Glass Baffles for Process Reactors, Chemglass
Stirrers Stirring Tools
Unique glass baffle system for process reactors from 2 to 50 L designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors
  • Available in square or rectangular shapes
  • Open or closed bottom
  • Baffle can be vertically adjusted inside reactor
PTFE joint is offset which permitting baffle to be adjusted up to 1" towards the center of the reactor. The effective baffle length for the reactor size 10 L to 20 L and for 30 L to 50 L are respectively 34 cm and 36 cm.
All baffles are designed with 3/4" precision ground topMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable