2-Octyl-1-dodecanol

Supplier: TCI America

O0429-25ML O0429-500ML
TCO0429-25MLEA 64.94 CAD
TCO0429-25ML TCO0429-500ML
2-Octyl-1-dodecanol
2-Octyl-1-dodecanol
Formula: C₂₀H₄₂O
MDL Number: MFCD01310428
CAS Number: 5333-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4714375cc28f1018da5510ab38d321667ac455ac&vid=4714375cc28f1018da5510ab38d321667ac455ac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR