PHCbi VIP® ECO Series Energy Efficient –86 °C Ultra-Low Temperature Upright Freezers, Energy Star Certified, PHC Corporation
Supplier: PHC Corporation
MDF-DU702VH-PA MDF-DU502VH-PA MDF-DU502VHA-PA MDF-DU702VHA-PA MDF-DU901VHA-PA MDF-DU502VH-PA MDF-DU702VHA-PA MDF-DU702VH-PA MDF-DU901VHA-PA MDF-DU502VHA-PA
76020-716EA 38290.19 CAD
76020-716 76269-676 76305-596 76305-598 76371-774 CA76269-676 CA76305-598 CA76020-716 CA76371-774 CA76305-596
PHCbi VIP® ECO Series Energy Efficient –86 °C Ultra-Low Temperature Upright Freezers, Energy Star Certified, PHC Corporation
Freezers Ultra Low Temperature Freezers
PHCbi's VIP® Series ultra-low temperature upright freezers are designed for safe, reliable storage of the most critical biologicals.
  • Reliable
  • Maximum storage
  • Minimal footprint
  • VIP Plus insulation composite
  • Uniform preservation
  • Microprocessor controls and alarms
  • Five-year manufacturer's parts and labor warranty
Energy efficiency in VIP Series freezers is achieved through balanced temperature performance and power management.
The advanced combination of cabinet design, electronics, refrigeration and critical components are engineered for enhancedMore Product Information