Puritan® PurSafe® DNA/RNA Preservative, Puritan Medical Products
Supplier: Puritan Medical Products
VL1201SAFE VL1201SAFE-H
CA76057-624BX 1070.02 CAD
CA76057-624 CA76057-626 CA76057-734 CA76057-736
Puritan® PurSafe® DNA/RNA Preservative, Puritan Medical Products
Vials
Intended for the stabilization and preservation of DNA and RNA of specimens to be analyzed by nucleic acids amplification tests.
  • Compatible with various sample types
  • Total microbial inactivation
  • Compatible with preservation of DNA/RNA from bacteria, viruses and tissues
  • HydraFlock® flocked swab maximizes sample collection and elution
  • For research use only
Simplify specimen collection and preservation for molecular research with PurSafe®.
Ordering information: Products are final sale.