Stirrer Shafts, Polished, Chemglass
Supplier: Chemglass
CG-2097-08 CG-2097-10 CG-2097-100 CG-2097-30 CG-2097-50 CG-2097-52 CG-2097-75 CG-2097-12 CG-2097-31 CG-2097-51 CG-2097-53 CG-2097-11 CG-2097-16
16003-324EA 783.45 CAD
16003-324 16003-326 16003-328 16003-330 16003-332 16003-334 16003-336 89029-050 89029-052 89029-054 89029-056 10170-262 10170-264
Stirrer Shafts, Polished, Chemglass
Stirrers Stirring Shafts
These precision ground and polished stirrer shaft are used with process reactors
The 16003-336 and 16003-328 are made from heavy wall tubing and have intends for agitator positioning, which are spaced for every 10" from the bottom of agitator.
Designed to utilize with PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8" wide flat.