2X YT Agar plates with Carbenicillin, Teknova

Supplier: TEKNOVA CA

Y4236 Y4391 Y4230 Y4390 Y4235 Y4238 Y4265 Y6260 Y6230
CA100217-086PK 1121.86 CAD
CA100217-086 CA100219-026 CA100219-364 CA100219-374 CA100219-792 CA101320-796 CA101320-800 CA101320-840 CA101446-776
2X YT Agar plates with Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology

These sterile pre poured agar plates are enriched with tryptone, yeast extract, sodium chloride, agar, glucose, carbenicillin


  • Shipping conditions: Ambient
  • Storage conditions: Refrigerate


They are designed for growth and propagation of recombinant E.coli and used with carbenicillin resistant strains.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR